missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin Antibody (182410), mFluor Violet 610 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1417MFV610
This item is not returnable.
View return policy
Description
Insulin Monoclonal antibody specifically detects Insulin in Human, Mouse, Bovine samples. It is validated for Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| Insulin | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Stem Cell Markers | |
| 3630 | |
| Store at 4C in the dark. |
| Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 182410 | |
| Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| E. coli-derived recombinant human Insulin | |
| RUO | |
| Primary | |
| Human, Mouse, Bovine | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction