missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin Antibody (D3E7 (5B6/6)), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64697AF405
This item is not returnable.
View return policy
Description
Insulin Monoclonal specifically detects Insulin in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Insulin | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| INS | |
| Recombinant human insulin. | |
| 0.1 mL | |
| Diabetes Research, Immune System Diseases, Immunology, Stem Cell Markers | |
| 3630 | |
| Human, Mouse, Rat, Pig, Bovine | |
| Purified |
| ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| D3E7 (5B6/6) | |
| ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| NB100-64697 recognises both insulin and proinsulin, but does not react with free C-peptide. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction