missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin Antibody (IRDN/805), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47727AF700
This item is not returnable.
View return policy
Description
Insulin Monoclonal antibody specifically detects Insulin in Human, Mouse, Porcine, Bovine, Rabbit, Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Insulin | |
| Monoclonal | |
| Alexa Fluor 700 | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| Recombinant Insulin protein (Uniprot: P01308) | |
| 0.1 mL | |
| Diabetes Research, Immune System Diseases, Immunology, Stem Cell Markers | |
| 3630 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| IRDN/805 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Pig, Bovine, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction