missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin Antibody (rIRDN/8546), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20705FR
This item is not returnable.
View return policy
Description
Insulin Monoclonal antibody specifically detects Insulin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Insulin | |
| Monoclonal | |
| DyLight 680 | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| Recombinant full-length human Insulin protein | |
| 0.1 mL | |
| Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Stem Cell Markers | |
| 3630 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Immunohistochemistry (Paraffin) | |
| rIRDN/8546 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction