missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin Antibody (SPM139), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34738MFV500
This item is not returnable.
View return policy
Description
Insulin Monoclonal antibody specifically detects Insulin in Human, Porcine, Bovine, Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry
Specifications
| Insulin | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Pig, Bovine, Mouse (Negative), Rat (Negative) | |
| Purified |
| Flow Cytometry | |
| SPM139 | |
| Flow Cytometry | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| Full length (1-84 amino acid) Purified pig insulin, conjugated to KLH | |
| 0.1 mL | |
| Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Stem Cell Markers | |
| 3630 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction