missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MafK Antibody (2F7), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22570AF350
This item is not returnable.
View return policy
Description
MafK Monoclonal specifically detects MafK in Human samples. It is validated for Western Blot, ELISA.
Specifications
| MafK | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| MAFK | |
| Recombinant human MAFK (1-156aa) purified from E. coli | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a κ |
| Western Blot, ELISA | |
| 2F7 | |
| Western Blot, ELISA | |
| FLJ32205, MGC71717, transcription factor MafK, ubiquitous (p18), v-maf avian musculoaponeurotic fibrosarcoma oncogene family, protein K, v-maf musculoaponeurotic fibrosarcoma (avian) oncogene family, protein K, v-maf musculoaponeurotic fibrosarcoma oncogene homolog K (avian) | |
| Mouse | |
| Protein A purified | |
| DNA replication Transcription Translation and Splicing, Epigenetics | |
| 7975.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction