missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin R/CD220 Antibody (243524) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1544JF525
This item is not returnable.
View return policy
Description
Insulin R/CD220 Monoclonal antibody specifically detects Insulin R/CD220 in Human samples. It is validated for Flow Cytometry,Immunocytochemistry
Specifications
| Insulin R/CD220 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR | |
| Mouse myeloma cell line NS0-derived recombinant human Insulin R/CD220, His28-Lys944, Accession # NP_001073285 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunocytochemistry | |
| 243524 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Diabetes Research, Lipid and Metabolism, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 3643 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction