Learn More
Insulysin/IDE Antibody (334501), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB2496CL1
Product Code. 30148311
  • € 986.00 / 0.10mL
Description
Insulysin/IDE Monoclonal antibody specifically detects Insulysin/IDE in Human samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Insulysin/IDE
Monoclonal
CoraFluor 1
Abeta-degrading protease, EC 3.4.24, EC 3.4.24.56, FLJ35968, Insulin protease, insulinase, insulin-degrading enzyme, Insulysin
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Insulysin/IDE, Met42-Leu1019, Accession # P14735
0.1 mL
Lipid and Metabolism
3416
Store at 4°C in the dark. Do not freeze.
IgG1
Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
334501
PBS
Mouse
Protein A or G purified