Learn More
Insulysin/IDE Antibody (334501), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2496CL1
This item is not returnable.
View return policy
Description
Insulysin/IDE Monoclonal antibody specifically detects Insulysin/IDE in Human samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| Insulysin/IDE | |
| Monoclonal | |
| CoraFluor 1 | |
| Abeta-degrading protease, EC 3.4.24, EC 3.4.24.56, FLJ35968, Insulin protease, insulinase, insulin-degrading enzyme, Insulysin | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Insulysin/IDE, Met42-Leu1019, Accession # P14735 | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 3416 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 334501 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |