missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulysin/IDE Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-61667
This item is not returnable.
View return policy
Description
Insulysin/IDE Polyclonal specifically detects Insulysin/IDE in Mouse, Rat samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| Insulysin/IDE | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| Abeta-degrading protease, EC 3.4.24, EC 3.4.24.56, FLJ35968, Insulin protease, insulinase, insulin-degrading enzyme, Insulysin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| Q8CGB9 | |
| IDE | |
| Peptide with sequence C-QQYNYDRDNIE corresponding to internal region according to NP_112419.2. | |
| 0.1 mg | |
| Lipid and Metabolism | |
| 3416 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction