missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 1/CD49a Antibody (CL7217) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76479F
This item is not returnable.
View return policy
Description
Integrin alpha 1/CD49a Monoclonal antibody specifically detects Integrin alpha 1/CD49a in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
Specifications
| Integrin alpha 1/CD49a | |
| Monoclonal | |
| FITC | |
| CD49 antigen-like family member A, CD49a, CD49a antigen, integrin, alpha 1, Laminin and collagen receptor, very late activation protein 1, VLA-1, VLA1integrin alpha-1 | |
| This antibody was developed using a synthetic peptide derived from P56199, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer | |
| 3672 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown | |
| CL7217 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction