missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 10 Antibody (885501) [FITC], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB9757F
This item is not returnable.
View return policy
Description
Integrin alpha 10 Monoclonal antibody specifically detects Integrin alpha 10 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Integrin alpha 10 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 885501 | |
| Flow Cytometry, CyTOF-ready | |
| integrin alpha-10, integrin, alpha 10, PRO827 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha 10 beta 1, Phe23-Thr1119 (Integrin alpha 10) and Gln21-Asp728 (Integrin beta 1), Accession # NP_001289400.1 (Integrin alpha 10) and P09055 (Integrin beta 1) | |
| 0.1 mL | |
| Signal Transduction | |
| 8515 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction