missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Integrin alpha 10 Antibody (885501) [PE/Cy7], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals™ FAB9757PECY7
This item is not returnable.
View return policy
Description
Integrin alpha 10 Monoclonal antibody specifically detects Integrin alpha 10 in Mouse samples. It is validated for Flow Cytometry
Specifications
| Integrin alpha 10 | |
| Monoclonal | |
| PE-Cyanine7 | |
| integrin alpha-10, integrin, alpha 10, PRO827 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha 10 beta 1, Phe23-Thr1119 (Integrin alpha 10) and Gln21-Asp728 (Integrin beta 1), Accession # NP_001289400.1 (Integrin alpha 10) and P09055 (Integrin beta 1) | |
| 0.1 mL | |
| Signal Transduction | |
| 8515 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry | |
| 885501 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction