missing translation for 'onlineSavingsMsg'
Learn More

Integrin alpha 2/CD49b Antibody (16B4) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

€ 201.00 - € 351.00

Specifications

Antigen Integrin alpha 2/CD49b
Clone 16B4
Concentration 1 mg/mL
Dilution Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18698977
View Documents
Novus Biologicals
NBP2-50144-0.025ml
0.025 mL
€ 201.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18650317
View Documents
Novus Biologicals
NBP2-50144
0.1 mL
€ 351.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Integrin alpha 2/CD49b Monoclonal antibody specifically detects Integrin alpha 2/CD49b in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, CyTOF
Specifications

Specifications

Integrin alpha 2/CD49b
1 mg/mL
Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Mouse
Cellular Markers, Innate Immunity, Signal Transduction
PBS
3673
IgG1
Protein A purified
16B4
Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready
Monoclonal
Purified
RUO
Human
alpha-2 subunit, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), VLA 2
Purified human beta 1 preparation from HT1080 fibrosarcoma cell extract.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.