missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 2/CD49b Antibody (235033), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1740CL1
This item is not returnable.
View return policy
Description
Integrin alpha 2/CD49b Monoclonal antibody specifically detects Integrin alpha 2/CD49b in Mouse samples. It is validated for Flow Cytometry,CyTOF
Specifications
| Integrin alpha 2/CD49b | |
| Monoclonal | |
| CoraFluor 1 | |
| alpha-2 subunit, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), VLA 2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Integrin alpha 2/CD49b, Tyr27-Thr1129, Accession # Q62469 | |
| 0.1 mL | |
| Cellular Markers, Innate Immunity, Signal Transduction | |
| 3673 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 235033 | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction