missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 2/CD49b Antibody (31H4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 201.00 - € 351.00
Specifications
| Antigen | Integrin alpha 2/CD49b |
|---|---|
| Clone | 31H4 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, CyTOF-ready |
| Applications | Flow Cytometry, ELISA, Immunoprecipitation, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18687237
|
Novus Biologicals
NBP2-50145-0.025ml |
0.025 mL |
€ 201.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18615728
|
Novus Biologicals
NBP2-50145 |
0.1 mL |
€ 351.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Integrin alpha 2/CD49b Monoclonal antibody specifically detects Integrin alpha 2/CD49b in Human samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, CyTOFSpecifications
| Integrin alpha 2/CD49b | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Immunoprecipitation, CyTOF | |
| Unconjugated | |
| Mouse | |
| Cellular Markers, Innate Immunity, Signal Transduction | |
| PBS | |
| 3673 | |
| IgG1 | |
| Protein A purified |
| 31H4 | |
| Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| alpha-2 subunit, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), VLA 2 | |
| Purified human b1 integrin preparation from HT1080 fibrosarcoma cell extract. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title