missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 2/CD49b Antibody (31H4), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50145DL594
This item is not returnable.
View return policy
Description
Integrin alpha 2/CD49b Monoclonal antibody specifically detects Integrin alpha 2/CD49b in Human samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, CyTOF
Specifications
| Integrin alpha 2/CD49b | |
| Monoclonal | |
| DyLight 594 | |
| alpha-2 subunit, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), VLA 2 | |
| Purified human b1 integrin preparation from HT1080 fibrosarcoma cell extract. | |
| 0.1 mL | |
| Cellular Markers, Innate Immunity, Signal Transduction | |
| 3673 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Immunoprecipitation, CyTOF | |
| 31H4 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido