Learn More
Integrin alpha 2/CD49b Antibody (CL7318) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76483B
Product Code. 30304501
  • € 737.00 / 0.10mL
Description
Integrin alpha 2/CD49b Monoclonal antibody specifically detects Integrin alpha 2/CD49b in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Integrin alpha 2/CD49b
Monoclonal
Biotin
alpha-2 subunit, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), VLA 2
This antibody was developed using a recombinant protein derived from P17301, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cellular Markers, Innate Immunity, Signal Transduction
3673
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL7318
PBS
Mouse
Protein A purified