missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD41/CD61 Chimeric Recombinant Rabbit Monoclonal Antibody (10E5)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547991
This item is not returnable.
View return policy
Description
Specificity: The antibody binds to GPIIb/IIIa with an affinity (Kd) of 16.2±6 nM. The antibody completely inhibits platelet aggregation at 7.8 μg/mL.
CD41 is a transmembrane glycoprotein (integrin family) composed of two chains GPIIb alpha (heavy chain; 120 kDa) and GPIIb beta (light chain; 23 kDa). CD61 non-covalently associates with CD41.
Specifications
| CD41/CD61 Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P05106, P08514 | |
| ITGA2B, ITGB3 | |
| Human blood platelets. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
| ELISA, Western Blot, Immunocytochemistry | |
| 10E5 | |
| Unconjugated | |
| ITGA2B | |
| alphaIIb protein; antigen CD61; BDPLT16; BDPLT2; CD41; CD41B; CD61; glycoprotein GPIIb; glycoprotein GPIIIa; glycoprotein IIb; glycoprotein IIIa; GP2B; GP3A; GPalpha IIb; GPIIb; GPIIIa; GT; GTA; HPA3; INTB3; integrin; integrin alpha 2b; integrin alpha 2b precursor; Integrin alpha-2b; integrin alpha-IIb; Integrin alpha-IIb heavy chain; Integrin alpha-IIb light chain, form 1; Integrin alpha-IIb light chain, form 2; Integrin beta; integrin beta 3; integrin beta 3 subunit; integrin beta3; integrin beta-3; integrin beta-3 subunit; integrin subunit alpha 2b; integrin subunit alpha IIb; integrin subunit beta 3; integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41); integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41B); integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); ITGA2B; ITGA2B-like protein; ITGAB; ITGB3; platelet fibrinogen receptor, alpha subunit; platelet glycoprotein IIb; platelet glycoprotein IIb of IIb/IIIa complex; platelet glycoprotein IIIa; platelet glycoprotein IIIa integrin subunit beta 3; platelet membrane glycoprotein IIb; Platelet membrane glycoprotein IIIa; platelet-specific antigen BAK; PPP1R93; protein phosphatase 1, regulatory subunit 93 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3674, 3690 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction