missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 2b/CD41 Antibody (MWReg30), Allophycocyanin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-43415APC
This item is not returnable.
View return policy
Description
Integrin alpha 2b/CD41 Monoclonal specifically detects Integrin alpha 2b/CD41 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Integrin alpha 2b/CD41 | |
| Monoclonal | |
| APC | |
| CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse | |
| IgG1 κ |
| Flow Cytometry | |
| MWReg30 | |
| Flow Cytometry | |
| ITGA2B | |
| The immunogen for this antibody was CD41. | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Immunology, Signal Transduction, Stem Cell Markers | |
| 3674 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction