missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 4 beta 1 Antibody (Hu114) [Janelia Fluor™ 585], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals FAB10603JF585
This item is not returnable.
View return policy
Description
Integrin alpha 4 beta 1 Monoclonal antibody specifically detects Integrin alpha 4 beta 1 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| Integrin alpha 4 beta 1 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| CD49d, IA4 | |
| Human Integrin alpha 4β1 | |
| RUO | |
| 3676 | |
| Store at 4°C in the dark. | |
| IgG4 |
| Flow Cytometry, CyTOF | |
| Hu114 | |
| 50mM Sodium Borate | |
| Human | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction