missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 4/CD49d Antibody (CL11294) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07999JF549
This item is not returnable.
View return policy
Description
Integrin alpha 4/CD49d Monoclonal antibody specifically detects Integrin alpha 4/CD49d in Human samples. It is validated for Western Blot
Specifications
| Integrin alpha 4/CD49d | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha | |
| This antibody was developed using a synthetic peptide derived from P13612, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones, Microautophagy, Neuronal Cell Markers, Phospho Specific | |
| 3676 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Western Blot | |
| CL11294 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction