missing translation for 'onlineSavingsMsg'
Learn More

Integrin alpha 5 beta 1 Antibody (SG/19) [Janelia Fluor« 646] - Chimeric, Novus Biologicals Biologicals™

Product Code. 30522539 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30522539 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30522539 Supplier Novus Biologicals Supplier No. NBP343579JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Integrin alpha 5 beta 1 Monoclonal antibody specifically detects Integrin alpha 5 beta 1 in Human samples. It is validated for Flow Cytometry,Immunoprecipitation,Functional Assay,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin alpha 5 beta 1
Applications Flow Cytometry, Immunoprecipitation, Functional Assay, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone SG/19
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias CD49 antigen-like family member E, CD49e, Fibronectin receptor subunit alpha, fibronectin receptor, alpha polypeptide, fibronectin receptor, alpha subunit, FNRA, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), ITGA5/SLC9B1 fusion, very late activation protein 5, alpha subunit, VLA-5, VLA5A
Host Species Rabbit
Immunogen The original antibody was generated by immunizing BALB/c mice with Nalm-6 cells. (Uniprot# P05556)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3678
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.