missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 5 beta 1 Antibody (SG/19), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43578
This item is not returnable.
View return policy
Description
Integrin alpha 5 beta 1 Monoclonal antibody specifically detects Integrin alpha 5 beta 1 in Human samples. It is validated for Flow Cytometry,Immunoprecipitation,Functional Assay,Immunocytochemistry/ Immunofluorescence
Specifications
| Integrin alpha 5 beta 1 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Functional | |
| CD49 antigen-like family member E, CD49e, Fibronectin receptor subunit alpha, fibronectin receptor, alpha polypeptide, fibronectin receptor, alpha subunit, FNRA, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), ITGA5/SLC9B1 fusion, very late activation protein 5, alpha subunit, VLA-5, VLA5A | |
| The original antibody was generated by immunizing BALB/c mice with Nalm-6 cells. (Uniprot# P05556) | |
| 0.1 mg | |
| Angiogenesis, Cancer, Immunology | |
| 3678 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Flow Cytometry, Immunoprecipitation, Functional Assay, Immunocytochemistry/Immunofluorescence | |
| SG/19 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction