missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-4 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41187UV
This item is not returnable.
View return policy
Description
Claudin-4 Polyclonal antibody specifically detects Claudin-4 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Claudin-4 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 350 | |
| claudin 4, Clostridium perfringens enterotoxin receptor, Clostridium perfringens enterotoxin receptor 1, CPE-RCPETR, CPE-receptor, hCPE-R, WBSCR8CPERCPETR1claudin-4, Williams-Beuren syndrome chromosomal region 8 protein | |
| Antibody was raised against a peptide corresponding to 17 amino acids near the carboxy terminus of human Claudin-4. The immunogen is located within the last 50 amino acids of Claudin-4. Amino Acid Sequence: PPRTDKPYSAKYSAAR | |
| 0.1 mL | |
| Cancer, Extracellular Matrix | |
| 1364 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction