missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 6/CD49f Antibody (GoH3), FITC, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-43400F
This item is not returnable.
View return policy
Description
Integrin alpha 6/CD49f Monoclonal antibody specifically detects Integrin alpha 6/CD49f in Human, Mouse, Bovine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
| Integrin alpha 6/CD49f | |
| Monoclonal | |
| FITC | |
| CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6B, VLA-6 | |
| The immunogen for this antibody was CD49f. | |
| 0.1 mL | |
| Signal Transduction | |
| 3655 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| GoH3 | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction