missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 6/CD49f Antibody (MP4F10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2676
This item is not returnable.
View return policy
Description
Integrin alpha 6/CD49f Monoclonal specifically detects Integrin alpha 6/CD49f in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, CyTOF-ready.
Specifications
| Integrin alpha 6/CD49f | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10 - 1:500, CyTOF-ready | |
| ITGA6 | |
| SW1222 human colorectal cell line. | |
| 0.1 mg | |
| Signal Transduction | |
| 3655 | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, CyTOF | |
| MP4F10 | |
| Unconjugated | |
| CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6B, VLA-6 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reacts with alpha 6. MP 4F10 can be used in clinical studies for tumour grading-stains moderately and well-differentiated colorectal adenocarcinoma. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction