missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 7 Antibody (334908) [Biotin], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB3518B
This item is not returnable.
View return policy
Description
Integrin alpha 7 Monoclonal antibody specifically detects Integrin alpha 7 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Integrin alpha 7 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 334908 | |
| Flow Cytometry, CyTOF-ready | |
| FLJ25220, integrin alpha 7 chain, integrin alpha-7, integrin, alpha 7 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha 7, Phe34-Pro1036, Accession # NP_032424 | |
| 0.1 mL | |
| Signal Transduction | |
| 3679 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction