missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 8 Antibody (481709) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6194B
This item is not returnable.
View return policy
Description
Integrin alpha 8 Monoclonal antibody specifically detects Integrin alpha 8 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| Integrin alpha 8 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8516 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 481709 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| integrin alpha-8, integrin, alpha 8 | |
| E. coli-derived recombinant human Integrin alpha 8, Glu610-Leu800, Accession # P53708 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction