missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha D/CD11d Mouse anti-Canine, DyLight 755, Clone: CA11.8H2, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65292IR-100UL
This item is not returnable.
View return policy
Description
Integrin alpha D/CD11d Monoclonal specifically detects Integrin alpha D/CD11d in Canine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| Integrin alpha D/CD11d | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ITGAD | |
| Affinity purified CD11 antigen from canine spleen | |
| 100 μL | |
| Signal Transduction | |
| 3681 | |
| Canine | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| CA11.8H2 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
| ADB2, CD11 antigen-like family member D, CD11D, FLJ39841, integrin alpha-D, integrin, alpha D, leukointegrin alpha D | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| NB100-65292 recognizes the canine CD11d cell surface antigen (the integrin ad sub-unit). The ad integrin is a newly recognized integrin chain that is expressed as a heterodimer with the b2 integrin (CD18). CD11d is expressed by a sub-population of CD8 positive T cells, macrophages and by gd T cells in the splenic red pulp. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction