Learn More
Integrin alpha E/CD103 Antibody (LF61), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-65272MFV450
Product Code. 18750561
  • € 608.00 / 0.10mL
Description
Integrin alpha E/CD103 Monoclonal antibody specifically detects Integrin alpha E/CD103 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Integrin alpha E/CD103
Monoclonal
mFluor Violet 450 SE
Hairy cell leukemia cells
Primary
Flow Cytometry
LF61
Mouse
Innate Immunity