Learn More
Integrin alpha E/CD103 Antibody (LF61), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65272MFV450
This item is not returnable.
View return policy
Description
Integrin alpha E/CD103 Monoclonal antibody specifically detects Integrin alpha E/CD103 in Human samples. It is validated for Flow Cytometry
Specifications
| Integrin alpha E/CD103 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| Hairy cell leukemia cells | |
| Primary |
| Flow Cytometry | |
| LF61 | |
| Mouse | |
| Innate Immunity |