missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha E2/Dendritic Cells Antibody (OX-62), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65618B
This item is not returnable.
View return policy
Description
Integrin alpha E2/Dendritic Cells Monoclonal antibody specifically detects Integrin alpha E2/Dendritic Cells in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Specifications
| Integrin alpha E2/Dendritic Cells | |
| Monoclonal | |
| Biotin | |
| antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen | |
| Density gradient enriched PVG rat thoracic duct dendritic cells. | |
| 0.25 mL | |
| Primary | |
| Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| OX-62 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 3682 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction