missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha L/CD11a Antibody (345913) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3595M
This item is not returnable.
View return policy
Description
Integrin alpha L/CD11a Monoclonal antibody specifically detects Integrin alpha L/CD11a in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, CyTOF
Specifications
| Integrin alpha L/CD11a | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Simple Western, Flow Cytometry, CyTOF | |
| 345913 | |
| Western Blot, Simple Western, Flow Cytometry, CyTOF-ready | |
| CD11 antigen-like family member A, CD11a antigen, CD11Aantigen CD11A (p180), lymphocyte function-associated antigen 1, alphapolypeptide, integrin alpha-L, integrin gene promoter, integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen1; alpha polypeptide), Leukocyte adhesion glycoprotein LFA-1 alpha chain, Leukocyte function-associated molecule 1 alpha chain, LFA-1, LFA-1 alpha, LFA1A, LFA-1A, lymphocyte function-associated antigen 1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Integrin alpha L, Tyr26-Met1089 (Tyr660Ile), Accession # CAA68747 | |
| 0.1 mL | |
| Adaptive Immunity, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers | |
| 3683 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction