Learn More
Integrin alpha L/CD11a Antibody (345913), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB3595MFV450
Product Code. 18528958
  • € 652.00 / 0.10mL
Description
Integrin alpha L/CD11a Monoclonal antibody specifically detects Integrin alpha L/CD11a in Human samples. It is validated for Western Blot, Western Blot, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Integrin alpha L/CD11a
Monoclonal
mFluor Violet 450 SE
50mM Sodium Borate
Mouse
0.1 mL
Adaptive Immunity, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers
3683
Store at 4C in the dark.
Western Blot, Flow Cytometry, CyTOF
345913
Western Blot, Simple Western, Flow Cytometry, CyTOF-ready
CD11 antigen-like family member A, CD11a antigen, CD11Aantigen CD11A (p180), lymphocyte function-associated antigen 1, alphapolypeptide, integrin alpha-L, integrin gene promoter, integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen1; alpha polypeptide), Leukocyte adhesion glycoprotein LFA-1 alpha chain, Leukocyte function-associated molecule 1 alpha chain, LFA-1, LFA-1 alpha, LFA1A, LFA-1A, lymphocyte function-associated antigen 1
Chinese hamster ovary cell line CHO-derived recombinant human Integrin alpha L, Tyr26-Met1089 (Tyr660Ile), Accession # CAA68747