missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha V beta 6 Antibody (7k), Alexa Fluor™ 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-50448AF594
This item is not returnable.
View return policy
Description
Integrin alpha V beta 6 Monoclonal specifically detects Integrin alpha V beta 6 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Integrin alpha V beta 6 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ITGAV | |
| 3T3b6, mouse fibroblast expressing human Integrin alpha V beta 6 | |
| Cancer, Cardiovascular Biology, Cellular Signaling, Stem Cells | |
| 3685 | |
| Human | |
| IgG2a |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 7k | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| CD51, integrin subunit alpha V, ITGAV, MSK8, VNRA, VTNR | |
| Rat | |
| 0.1 mL | |
| Primary | |
| Does not bind mouse. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction