Learn More
Integrin alpha V beta 6 Antibody (7k), mFluor Violet 610 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-50448MFV610
This item is not returnable.
View return policy
Description
Integrin alpha V beta 6 Monoclonal antibody specifically detects Integrin alpha V beta 6 in Human samples. It is validated for Flow Cytometry
Specifications
| Integrin alpha V beta 6 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Cancer, Cardiovascular Biology, Cellular Signaling, Stem Cells | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| 7k | |
| Flow Cytometry | |
| CD51, integrin subunit alpha V, ITGAV, MSK8, VNRA, VTNR | |
| Human | |
| Purified |