Learn More
Integrin alpha V beta 6 Antibody (STX-100), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-28326CL1
Product Code. 30149854
  • € 879.00 / 0.10mL
Description
Integrin alpha V beta 6 Monoclonal antibody specifically detects Integrin alpha V beta 6 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Integrin alpha V beta 6
Monoclonal
CoraFluor 1
CD51, integrin subunit alpha V, ITGAV, MSK8, VNRA, VTNR
Integrin aVb6 (ITGAV & ITGB6)
0.1 mL
Cancer, Cardiovascular Biology, Cellular Signaling, Stem Cells
3685
Store at 4°C in the dark. Do not freeze.
IgG1
Flow Cytometry, ELISA, Functional Assay
STX-100
PBS
Human
Protein A purified