Learn More
Novus Biologicals™ Integrin alpha V beta 6 Antibody (STX-100) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28326-100ug
 View more versions of this product
Product Code. 30048545
  • € 349.00 / 100µg
Description
Integrin alpha V beta 6 Monoclonal antibody specifically detects Integrin alpha V beta 6 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Integrin alpha V beta 6
Monoclonal
Unconjugated
CD51, integrin subunit alpha V, ITGAV, MSK8, VNRA, VTNR
Integrin aVb6 (ITGAV & ITGB6)
100 μg
Cancer, Cardiovascular Biology, Cellular Signaling, Stem Cells
3685
Human
Purified
Flow Cytometry, ELISA, Functional Assay
STX-100
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.