missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha V/CD51 Antibody (23C6), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08850AF405
This item is not returnable.
View return policy
Description
Integrin alpha V/CD51 Monoclonal antibody specifically detects Integrin alpha V/CD51 in Human, Chicken, Bovine (Negative), Rabbit (Negative) samples. It is validated for ELISA
Specifications
| Integrin alpha V/CD51 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| antigen identified by monoclonal L230, CD51, CD51 antigen, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), MSK8, Vitronectin receptor subunit alpha, VNRADKFZp686A08142 | |
| Osteoclasts from osteoclastomas | |
| 100 μg | |
| Adaptive Immunity, Angiogenesis, Cancer, Cell Biology, Cellular Markers, Immunology, Signal Transduction | |
| 3685 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA | |
| 23C6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Chicken, Bovine (Negative), Rabbit (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction