missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha V/CD51 Mouse anti-Human, Clone: ITGAV/1610, Novus Biologicals™
Mouse Monoclonal Antibody
€ 508.00
Specifications
| Antigen | Integrin alpha V/CD51 |
|---|---|
| Clone | ITGAV/1610 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, ELISA 1-5 μg/mL, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunofluorescence |
Description
Integrin alpha V/CD51 Monoclonal specifically detects Integrin alpha V/CD51 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Integrin alpha V/CD51 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Angiogenesis, Cancer, Cell Biology, Cellular Markers, Immunology, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 3685 | |
| IgG1 κ | |
| Protein A or G purified |
| ITGAV/1610 | |
| Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, ELISA 1-5 μg/mL, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| antigen identified by monoclonal L230, CD51, CD51 antigen, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), MSK8, Vitronectin receptor subunit alpha, VNRADKFZp686A08142 | |
| Recombinant human full-length Integrin alpha V/CD51 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title