missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 1/CD29 Antibody (3B6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
€ 210.00
Specifications
| Antigen | Integrin beta 1/CD29 |
|---|---|
| Clone | 3B6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
Description
Integrin beta 1/CD29 Monoclonal specifically detects Integrin beta 1/CD29 in Human, Porcine, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| Integrin beta 1/CD29 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| RUO | |
| Human, Pig, Monkey, Primate | |
| 3688 | |
| Purified recombinant fragment of human Integrin beta 1 expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 3B6 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Cancer, Cytokine Research, Cytoskeleton Markers, Embryonic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA | |
| ITGB1 | |
| IgG1 | |
| Affinity Purified | |
| 88.4 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title