missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 1/CD29 Antibody (MEM-101A), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-383H
This item is not returnable.
View return policy
Description
Integrin beta 1/CD29 Monoclonal antibody specifically detects Integrin beta 1/CD29 in Human, Porcine, Canine, Mouse (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF
Specifications
| Integrin beta 1/CD29 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Pig, Canine, Mouse (Negative) | |
| Purified |
| Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF | |
| MEM-101A | |
| Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF-ready | |
| CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA | |
| Raji Burkitt's lymphoma cell line (NM_002211.3). | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| 3688 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction