missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 1/CD29 Antibody (MEM-101A), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-383PEATT594
This item is not returnable.
View return policy
Description
Integrin beta 1/CD29 Monoclonal specifically detects Integrin beta 1/CD29 in Human, Porcine, Canine, Mouse (Negative) samples. It is validated for Flow Cytometry.
Specifications
| Integrin beta 1/CD29 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| ITGB1 | |
| Raji Burkitt′s lymphoma cell line Raji (NM_002211.3). | |
| 0.1 mL | |
| Primary | |
| The antibody MEM-101A reacts with CD29 (integrin beta1 chain), a 130 kDa single chain type I glycoprotein expressed as a heterodimer (non-covalently associated with the integrin alpha subunits 1-6). CD29 is broadly expressed on majority of hematopoietic and non-hematopoietic cells (leukocytes, platelets, fibroblasts, endothelial cells, epithelial cells and mast cells). HLDA VI; WS Code AS A048 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| MEM-101A | |
| Flow Cytometry | |
| CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA | |
| Mouse | |
| Protein A purified | |
| Cancer, Cytokine Research, Cytoskeleton Markers, Embryonic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| 3688.0 | |
| Human, Pig, Canine, Mouse (Negative) | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction