missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 1D Antibody (2B1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-97734CL1
This item is not returnable.
View return policy
Description
Integrin beta 1D Monoclonal antibody specifically detects Integrin beta 1D in Canine,Human,Mouse,Porcine samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Frozen),Immunocytochemistry/Immunofluorescence
Specifications
| Integrin beta 1D | |
| Monoclonal | |
| CoraFluor 1 | |
| integrin beta-1, CD29, FNRB, GPIIA, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12), MDF2, MSK12, VLAB, VLA-BETA | |
| Derived by fusion of SP2/0 Mouse myeloma cells with spleen cells from a Mouse immunized with a synthetic peptide corresponding to the C-terminal 24 amino acids of Integrin beta 1D including an appending N-terminal cysteine (CQENPIYKS-PINNFKNPNYGRKAGL) coupled to keyhole limpet hemocyanin. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| 3688 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence | |
| 2B1 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Canine, Human, Mouse, Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction