missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 2/CD18 Antibody (MEM-148), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-480FR
This item is not returnable.
View return policy
Description
Integrin beta 2/CD18 Monoclonal specifically detects Integrin beta 2/CD18 in Human, Porcine (Negative) samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| Integrin beta 2/CD18 | |
| Monoclonal | |
| DyLight 680 | |
| P05107 | |
| ITGB2 | |
| Human tissue cell preparation, Peripheral blood mononuclear cells (NM_000211.3). | |
| 0.1 mL | |
| Apoptosis, Cytokine Research, Immunology, Phospho Specific | |
| 3689 | |
| Human, Porcine (Negative) | |
| Purified |
| Western Blot, Flow Cytometry | |
| MEM-148 | |
| Western Blot, Flow Cytometry | |
| 959 beta subunit precursor)10, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, MF17, MFI7, P150 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Recognizes the epitope exposed on free human CD18 chains unassociated with CD11 as well as on high-affinity state of LFA-1. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction