missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 2/CD18 Antibody (MEM48), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14608CL1
This item is not returnable.
View return policy
Description
Integrin beta 2/CD18 Monoclonal antibody specifically detects Integrin beta 2/CD18 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot,Functional Assay
Specifications
| Integrin beta 2/CD18 | |
| Monoclonal | |
| CoraFluor 1 | |
| 959 beta subunit precursor)10, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, MF17, MFI7, P150 | |
| Leucocytes from an LGL - type leukaemia | |
| 0.1 mL | |
| Apoptosis, Cytokine Research, Immunology, Phospho Specific | |
| 3689 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Functional Assay | |
| MEM48 | |
| PBS | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction