Learn More
Integrin beta 3/CD61 Antibody (256809) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB2266JF525
Product Code. 30169824
  • € 617.00 / 0.10mL
Description
Integrin beta 3/CD61 Monoclonal antibody specifically detects Integrin beta 3/CD61 in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Integrin beta 3/CD61
Monoclonal
Janelia Fluor 525
CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa
Chinese hamster ovary cell line CHO-derived recombinant human Integrin β3/CD61, Gly27-Asp718, Accession # P05106
0.1 mL
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immunology, Phospho Specific, Signal Transduction, Stem Cell Markers
3690
Store at 4°C in the dark.
IgG2a
Flow Cytometry, Western Blot, CyTOF
256809
50mM Sodium Borate
Mouse
Protein A or G purified