missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 3/CD61 Antibody (CL7319) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76484PE
This item is not returnable.
View return policy
Description
Integrin beta 3/CD61 Monoclonal antibody specifically detects Integrin beta 3/CD61 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Integrin beta 3/CD61 | |
| Monoclonal | |
| PE | |
| CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa | |
| This antibody was developed using a recombinant protein derived from P05106, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immunology, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| 3690 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL7319 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction