missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 3/CD61 Antibody (VIPL2), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
€ 577.00
Specifications
| Antigen | Integrin beta 3/CD61 |
|---|---|
| Clone | VIPL2 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), CyTOF |
| Classification | Monoclonal |
| Conjugate | Biotin |
Description
Integrin beta 3/CD61 Monoclonal antibody specifically detects Integrin beta 3/CD61 in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), CyTOFSpecifications
| Integrin beta 3/CD61 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), CyTOF | |
| Biotin | |
| Mouse | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immunology, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| PBS | |
| 3690 | |
| IgG1 κ | |
| Protein A purified |
| VIPL2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Primate | |
| CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa | |
| The mouse monoclonal antibody VIPL2 recognizes CD61 | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title