missing translation for 'onlineSavingsMsg'
Learn More
Learn More
integrin beta 4 binding protein Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-52640B
This item is not returnable.
View return policy
Description
integrin beta 4 binding protein Polyclonal antibody specifically detects integrin beta 4 binding protein in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| integrin beta 4 binding protein | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| b(2)gcn, B(2)GCN homolog, B4 integrin interactor, CAB, EIF3Ap27(BBP), eIF-6, eukaryotic translation initiation factor 3A, eukaryotic translation initiation factor 6, ITGB4BPintegrin beta 4 binding protein, p27 beta-4 integrin-binding protein, p27BBP | |
| The immunogen for this product maps to a region between residue 50 and 100 of human Eukaryotic Translation Initiation Factor 6 using the numbering given in entry NP_852133.1 (GeneID 3692). | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 3692 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction