missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 4/CD104 Antibody (450-9D), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65599C
This item is not returnable.
View return policy
Description
Integrin beta 4/CD104 Monoclonal specifically detects Integrin beta 4/CD104 in Human, Ferret, Mustelid samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| Integrin beta 4/CD104 | |
| Monoclonal | |
| DyLight 650 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ITGB4 | |
| Purified alpha6 beta4 integrin from A431 cells | |
| 0.1 mL | |
| Primary | |
| NB100-65599 recognizes the human beta4 integrin (also known as CD104), a 205kD glycoprotein which associates with the alpha6 integrin to form the alpha6/beta4 complex. CD104 is expressed on epithelial cells, Schwann cells and various tumour cell lines. NB100-65599 recognizes an extracellular epitope on the CD104 molecule. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| 450-9D | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| CD104, CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, integrin, beta 4 | |
| Mouse | |
| Protein G purified | |
| Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
| 3691.0 | |
| Human, Ferret, Mustelid | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction